Tubes, both ends open, ceramic
Toimittaja: Thermo Fisher Scientific
31554.DW 31558.DM 31558.DW 31558.EO 31563.EH 31563.EM 31572.DW 31572.EH 31573.DR 31573.DW 31573.EH 31574.DR 31574.EH 31575.DM 31575.DW 31575.EO
31554.DWEA 161 EUR
31554.DW 31558.DM 31558.DW 31558.EO 31563.EH 31563.EM 31572.DW 31572.EH 31573.DR 31573.DW 31573.EH 31574.DR 31574.EH 31575.DM 31575.DW 31575.EO
Tubes, both ends open, ceramic
Letku Siirtoputki
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.