Insulating tubes, ceramic
Toimittaja: Thermo Fisher Scientific
31475.DR 31476.DR 31476.DT 31479.DR 31479.DW 31492.DR 31475.DT 31475.DW 31485.DR 31485.DW 31487.DR 31487.DW
31475.DREA 75.7 EUR
31475.DR 31476.DR 31476.DT 31479.DR 31479.DW 31492.DR 31475.DT 31475.DW 31485.DR 31485.DW 31487.DR 31487.DW
Insulating tubes, ceramic
Letku Siirtoputki
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.