Methylene Violet 3RAX 90% (dye content), Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ100

307505-1G 307505-5G
307505-1GEA 85.9 EUR
307505-1G 307505-5G
Methylene Violet 3RAX 90% (dye content), Sigma-Aldrich®
Methylene Violet 3RAX

Methylene Violet 3RAX has been used as a reagent in a preparation of citrate methylene violet, which is used for staining yeast cells. It is also used as an internal standard in tandem mass spectrometry (MS/MS) for the quantification of methylene blue in blood sample.


Methylene violet 3RAX [MV3RAX, 3-amino-7-(diethylamino)-5-phenyl phenazinium chloride] is a phenazinium dye and is cationic in nature. It has several applications in staining chemistry. Methylene violet 3RAX dye has an antimalarial property. It is used as a probe for investigating guest–host interactions in dendrimers and this may have relevance in drug–DNA interactions.

Kaava: C₂₂H₂₃ClN₄
Sulamispiste: 285 °C (haj. lämpötila, lit.)
Storage Temperature: Room Temperature
MDL numero: MFCD00013433
CAS-numero: 4569-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01cf4373a3807b6a817cc807b972d8dc2389a227&vid=8c2c5a2c97f6a308ce18422c96f39c1fa820a83c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance (Colour) Green to Very Dark Green
Appearance (Form) Crystalline Powder
Infrared Spectrum Conforms to Structure
Wavelength 231 - 237 nm
Extinction Coefficient ≥12000
Wavelength 271 - 277 nm
Purity (HPLC) Typically ≥90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR