Solvent blue 35, violet crystalline powder, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

306436-100G 306436-5G
306436-100GEA 209 EUR
306436-100G 306436-5G
Solvent blue 35, violet crystalline powder, Sigma-Aldrich®
Solvent blue 35

Synonyms: Solvent blue 35

Kaava: C₂₂H₂₆N₂O₂
Sulamispiste: 120…122 °C (kirj.)
MDL numero: MFCD00011714
CAS-numero: 17354-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b127fb1a7d0230741a67989ac722c4979a5745&vid=21b127fb1a7d0230741a67989ac722c4979a5745&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Composition Dye content, 98%
Absorption λmax 604 nm (2nd)
Appearance (color) Maroon to violet or purple
Appearance (form) Powder or crystalline powder
Infrared spectrum Conforms to structure
Wavelength, c = 0.01g/l in chloroform 648 - 652 nm
Extinction coefficient ≥13000
Wavelength 600 - 604 nm
Carbon 73,5 - 77,3%
Nitrogen 7,7 - 8,3%
Solubility (turbidity), c = 4 mg/ml in chloroform, clear to opaque Conforms
Solubility (color) Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR