Bis(2,4-pentanedionato)tin(IV) dichloride ≥95%

Toimittaja: Thermo Fisher Scientific
Vaara

30532.09 30532.18
30532.09EA 220 EUR
30532.09 30532.18
Bis(2,4-pentanedionato)tin(IV) dichloride ≥95%
Bis(2,4-pentanedionato)tin(IV) dichloride

Purity: 95% CAS No.: 16919-46-3 Molecular Formula: SnCl2(C5H7O2)2 Formula Weight: 387.83 Physical Form: Crystalline Sensitivity: Moisture Sensitive Melting Point: 199° UN No.: UN3146 MDL No.: MFCD00015316

Kaava: C₁₀H₁₄Cl₂O₄Sn
Sulamispiste: 199 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00015316
CAS-numero: 16919-46-3
EINECS: 240-972-5
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR