Diammoniumvetyortofosfaatti ≥98% (by alkalimetry), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Toimittaja: Honeywell Chemicals

30402-50KG 30402-1KG 30402-500G
30402-50KGEA 1900 EUR
30402-50KG 30402-1KG 30402-500G
Diammoniumvetyortofosfaatti ≥98% (by alkalimetry), Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Diammoniumvetyortofosfaatti
Kaava: (NH₄)₂HPO₄
Sulamispiste: 155 °C
Tiheys: 1,62 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010891
CAS-numero: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now

Testitulosten erittely

Assay Min. 98.0 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 7.7 - 8.1
Calcium (Ca) Max. 0.001 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.0005 %
Sodium (Na) Max. 0.005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.001 %
Nitrate (NO₃) Max. 0.003 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR