Ceramic tubes, one end closed
Toimittaja: Thermo Fisher Scientific
30062.DR 30062.DW 30062.EH 30062.EM 30062.EO 30073.DR 30080.DM 30080.DW 30080.EH
30062.DREA 210 EUR
30062.DR 30062.DW 30062.EH 30062.EM 30062.EO 30073.DR 30080.DM 30080.DW 30080.EH
Ceramic tubes, one end closed
Letku Siirtoputki
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.