Human plasma, defibrinated, delipidated

Toimittaja: SEQENS

30010-100ML 30010-500ML 30010-1L
30010-100MLEA 180 EUR
30010-100ML 30010-500ML 30010-1L
Human plasma, defibrinated, delipidated
Plasma

SEQENS IVD manufactures Defibrinated Delipidated Human Plasma. It is dialysed and electrolytes balanced.


  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • Fibrinogen: Absent
  • Cholesterol: <0,04 g/L
  • Triglycerides: <0,05 g/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years


For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.


Complete donor traceability on file.


Varoitus: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR