Dipping bottles
Toimittaja: Sampling Systems
5332B-1000 5332B-300 5332B-500 5332S-1000 5332S-300 5332S-500
300-0482EA 440 EUR
300-0482 300-0483 300-0484 300-0485 300-0486 300-0487
Dipping bottles
Näytteenottimet Nestenäytteet
Dipping bottles are excellent for sampling in open water, as well as in tanks and drums.
  • Wide range available
  • Stainless steel 304 or brass construction
  • Customisable
Sampling liquids at depth is easy with these dipping bottles. Available in either 304 stainless steel or brass construction, they can take liquid samples from a variety of locations including open water kegs, drums, and wells.