Methyl cellulose, 300 - 560 mPa.s, Methocel® A4C

Toimittaja: VWR Chemicals

Synonyms: Cellulose methyl ether

298944NEA 257 EUR
298944N
Methyl cellulose, 300 - 560 mPa.s, Methocel® A4C
Methyl cellulose
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081763
CAS-numero: 9004-67-5

Order Now

Testitulosten erittely

Viscosity 300 - 560 mPa.s
pH (1 %) 5.0 - 8.0
Heavy metals (as Pb) Max. 0.001 %
Loss on drying Max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR