Aluminium foils rolls and dispenser
Toimittaja: Korff
60010 60050 60020 60060 60004 60120 60130 60150 60012 60100 60170 60175 60110 60168 60171 60172 60187 60179 60009 60140 64044 64046 60195 60180 60160 60165 60166 60185
293-4111EA 15.1 EUR
293-4111 293-4110 293-4112 293-4113 293-4201 291-0009 291-0010 291-0011 KORF60012 611-9036 291-0042 291-0043 KORF60110 KORF60168 KORF60171 KORF60172 KORF60187 KORF60179 KORF60009 KORF60140 KORF64044 KORF64046 KORF60195 KORF60180 KORF60160 KORF60165 KORF60166 KORF60185
Aluminium foils rolls and dispenser
Alumiinifoliot
Pure aluminium, 99,0 to 99,5%, soft.
  • Different widths and thicknesses available
  • Comes in a practical dispenser
  • Economical large size rolls for use in a roll dispenser
Ideal for packaging, covering and insulation.