Adhesive PCR sealing film and foil seals
Toimittaja: Simport Scientific
T329-1 T329-2 T329-5
293-1401EA 110 EUR
293-1401 293-1402 293-1405
Adhesive PCR sealing film and foil seals
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Transparent adhesive sealing film is ideal for reducing cross contamination and/or spill over in PCR applications. 
  • Thermostable and functional from −70 to +95 °C
  • Choice of sterile or non sterile
Aluminium sealing foil is suitable for both manual sealing during PCR work and high throughput applications. Pierceable with a pipette tip for easy access to sample. Adhesive backing makes it easy to apply. Will resist temperatures from –80 to +120 °C. It is recommended that the foil be applied using the specially designed rubber roller to ensure aMore Product Information