Size exclusion chromatography columns, Superdex™ 30 Increase
Toimittaja: Cytiva
29-2197-57 29-2197-58
29-2197-57EA 2865 EUR
29-2197-57 29-2197-58
Size exclusion chromatography columns, Superdex™ 30 Increase
Kromatografiakolonnit
Superdex™ 30 Increase columns provide high resolution separations under mild conditions for small biomolecules having a molecular weight between 100 and 7000, such as peptides and oligosaccharides.
  • Runtime reduced down to one third with maintained resolution for faster results
  • Up to 50% higher resolution for improved purity and analysis results
  • Higher lot-to-lot consistency