Trietyyliamiini ≥99.6%, HiPerSolv CHROMANORM® for HPLC

Toimittaja: VWR Chemicals
Vaara

Synonyms: (Diethylamino)ethane, TEA

Filtered through a 0.2 µm filter, packaged under nitrogen.

28757.184EA 74.7 EUR
28757.184 28757.230
Trietyyliamiini ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Trietyyliamiini
Kaava: (C₂H₅)₃N
MW: 101.19 g/mol
Kiehumispiste: 90
Sulamispiste: –115 °C
Tiheys: 0.729 g/cm³ (20 °C)
Leimahduspiste: –11 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009051
CAS-numero: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 (8) II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99.6 %
IR Spectrum Passes test
Refractive index (20°C) 1.399 - 1.401
Evaporation residue Max. 50 ppm
Water Max. 0.2 %
Fe (Iron) Max. 0.2 ppm
Pb (Lead) Max. 0.02 ppm
Transmittance (254 nm) Min. 20 %
Transmittance (280 nm) Min. 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR