Trietanoliamiini ≥99.0% analytical reagent

Toimittaja: VWR Chemicals

28746.290EA 113 EUR
28746.290 28746.320
Trietanoliamiini ≥99.0% analytical reagent
Trietanoliamiini
Kaava: C₆H₁₅NO₃
Kiehumispiste: 335,4 °C (1013 hPa)
Tiheys: 1,13 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002855
CAS-numero: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay Min. 99.0 %
IR Spectrum Passes test
Density (20/4) 1.120 - 1.130
Solidification point 20 - 22 °C
Diethanolamine Max. 0.5 %
Ethanolamine Max. 0.1 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 50 ppm
Water Max. 0.2 %
Fe (Iron) Max. 10 ppm

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR