Tioasetamidi ≥99.0% analytical reagent

Toimittaja: VWR Chemicals
Vaara

Synonyms: Ethanethioamide

28603.137EA 42.7 EUR
28603.137
Tioasetamidi ≥99.0% analytical reagent
Tioasetamidi
Kaava: CH₃CSNH₂
MW: 75.13 g/mol
Sulamispiste: 113…114 °C
Tiheys: 1.336 g/cm³ (20 °C)
MDL numero: MFCD00008070
CAS-numero: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay Min. 99.0 %
Melting point 110 - 113 °C
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.05 %
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR