Tetrahydrofuraani ≥99.8%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials
Vaara

2858-25EA 293.21 EUR
2858-25
Tetrahydrofuraani ≥99.8%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Tetrahydrofuraani
Kaava: C₄H₈O
Kiehumispiste: 65 °C (101,325 kPa)
Sulamispiste: –108,3 °C
Tiheys: 0,888 g/cm³ (20 °C)
Leimahduspiste: –14,00 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005356
CAS-numero: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use ≥ 99.8 %
Assay (C₄H₈O) (by GC, corrected for water) ≥99.8 %
Peroxide (as H₂O₂) ≤0.015 %
Residue after Evaporation ≤5 ppm
Water (H₂O)(by KF, coulometric) ≤0.03 %
Optical Absorbance (1-cm path vs water) - 212 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 225 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 250 nm ≤0.17
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.01
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR