Lisävarusteet, GC septa, Thermogreen® LB-2
Toimittaja: Merck
20654 20652 23159 23154 20651 20666 23156 23157 28021-U
SUPL20654EA 166 EUR
SUPL20654 20652. 23159. SIAL23154 20651. SUPL20666 23156. 23157. 28021-U
Lisävarusteet, GC septa, Thermogreen® LB-2
Septumi
Ready to use GC septa for inlet temperature of up to 100 to 350 °C.
  • Low bleeding
  • Easy needle penetration
  • High puncture tolerance and ideal for autosamplers