Affinity chromatography column, GSTPrep™ FF
Toimittaja: Cytiva
28-9365-50
28-9365-50EA 963 EUR
28-9365-50
Affinity chromatography column, GSTPrep™ FF
Kromatografiakolonnit
GSTPrep™ FF 16/10 is prepacked, ready-to-use column containing glutathione Sepharose™ 4 fast flow designed for one-step, scale-up purification of glutathione S-transferase (GST)-tagged proteins.
  • Binds approximately 200 mg recombinant GST/column
  • Mild elution conditions preserving protein antigenicity and function
  • Simple operation with a pump or chromatography system, such as ÄKTA design
Glutathione Sepharose™ 4 fast flow is for scale-up purification of recombinant GST-tagged proteins, other glutathione S-transferases, and glutathione-dependent proteins.More Product Information