Gel filtration columns, HiPrep™ with Sephacryl® S-400 HR
Toimittaja: Cytiva
28-9356-04 28-9356-05
28-9356-04EA 865 EUR
28-9356-04 28-9356-05
Gel filtration columns, HiPrep™ with Sephacryl® S-400 HR
Kromatografiakolonnit
HiPrep™ 16/60 Sephacryl® S-400 HR and HiPrep™ 26/60 Sephacryl® S-400 HR are pre-packed gel filtration columns for preparative separation of biomolecules of up to 5 and 13 ml sample volume respectively. Sephacryl® S-400 is used for separating polysaccarides, macromolecules with extended structures, and even small plasmids.
  • Maximum recovery and minimum non specific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Columns provides reproducible separations over a wide molecular weight range