Suodattimet, näytekonsentraattorit, Vivaspin®
Toimittaja: Cytiva
28-9322-40 28-9322-37 28-9322-36 28-9322-35 28-9322-25 28-9322-23 28-9322-18 28-9322-47 28-9322-48 28-9323-61 28-9322-45 28-9323-62 28-9323-60 28-9323-59 28-9323-58 28-9323-19 28-9323-18 28-9323-17 28-9322-96 28-9322-94 28-9322-93 28-9322-58 28-9322-57 28-9323-63
28-9322-40EA 216 EUR
28-9322-40 28-9322-37 28-9322-36 28-9322-35 28-9322-25 28-9322-23 28-9322-18 28-9322-47 28-9322-48 28-9323-61 28-9322-45 28-9323-62 28-9323-60 28-9323-59 28-9323-58 28-9323-19 28-9323-18 28-9323-17 28-9322-96 28-9322-94 28-9322-93 28-9322-58 28-9322-57 28-9323-63
Suodattimet, näytekonsentraattorit, Vivaspin®
Suodattimet Sentrifugisuodattimet
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
  • Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • Vertical PES membrane minimises membrane blockage and tolerates high flow rates
  • Compatible pH range is from pH 1 to 9
Up to 30-fold concentration of the sample can be achieved with recovery ofMore Product Information