Disposable columns prepacked, Sepharose™ His SpinTrap™ kit
Toimittaja: Cytiva
28-9321-71
28-9321-71EA 475 EUR
28-9321-71
Disposable columns prepacked, Sepharose™ His SpinTrap™ kit
Kromatografiakolonnit
His SpinTrap™ is a prepacked, single use spin column for purifying histidine-tagged proteins by immobilised metal ion affinity chromatography (IMAC) and rapid expression screening.
  • Ni Sepharose™ High Performance in prepacked spin columns
  • Yields up to 750 µg pure protein per column
  • Allows direct purification of both clarified and unclarified cell lysates
Toimitustiedot: Kit contains one pack of His SpinTrap™ and one pack of His buffer kit.