Natriumasetaatti ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Toimittaja: VWR Chemicals

Synonyms: Acetic acid sodium salt

27653.235EA 37 EUR
27653.235 27653.292 27653.260 27653.460 27653.360
Natriumasetaatti ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Natriumasetaatti
Kaava: H₃CCOONa
MW: 82.03 g/mol
Kiehumispiste: 881.4 °C (1013 hPa)
Sulamispiste: 324
Tiheys: 1.528 g/cm³ (20 °C)
Leimahduspiste: >250 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012459
CAS-numero: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Testitulosten erittely

Assay Min. 99.0 %
Identification Passes test
IR Spectrum Passes test
pH (25°C; 5 %) 7.0 - 9.2
Heavy metals (as Pb) Max. 10 ppm
Insolubility in water Max. 100 ppm
Loss on drying (120°C) Max. 1.0 %
Cl (Chloride) Max. 20 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 30 ppm
Al (Aluminium) Max. 10 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 3 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 0.05 %
Mg (Magnesium) Max. 20 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR