Etikkahappo 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP

Toimittaja: Honeywell Chemicals
Vaara

27221-1L 27221-2.5L 27221-5L 27221-4X2.5L 27221-4X5L 27221-6X1L
27221-1LEA 62.9 EUR
27221-1L 27221-2.5L 27221-5L 27221-4X2.5L 27221-4X5L 27221-6X1L
Etikkahappo 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP
Etikkahappo
Kaava: H₃CCOOH
Kiehumispiste: 118 °C (1,013 hPa)
Sulamispiste: 16 °C
Tiheys: 1,05 g/cm³ (20 °C)
Leimahduspiste: 40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay Min. 99 %
Non-volatile matter Max. 0.005 %
Iron (Fe) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Sulfate (SO₄) Max. 0.0005 %
Explosion Limit 4 %, 59 °F
Vapor Density 2.07 (vs air)
Autoignition Temperature 800 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR