Lyijy(II,IV)oksidi, red, TECHNICAL

Toimittaja: VWR Chemicals
Vaara

26572.295EA 105 EUR
26572.295
Lyijy(II,IV)oksidi, red, TECHNICAL
Lyijy(II,IV)oksidi
Kaava: Pb₃O₄
Kiehumispiste: 2682 °C (1013 hPa)
Sulamispiste: 830 °C
Tiheys: 9,1 g/cm³ (20 °C)
MDL numero: MFCD00078491
CAS-numero: 1314-41-6
EINECS: 215-235-6
UN: 1479
ADR: 5.1,II
Merck Index: 12,05449

Order Now

Testitulosten erittely

Assay (calculated as PbO2) Min. 30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR