Inlet liners, SPME-GC, Supelco®
Toimittaja: Merck
2636425 2637501 2637505 2637525 2633901 2633905 2635825
2636425.EA 542 EUR
2636425. 2637501. 2637505. 2637525. 2633901. 2633905. 2635825.
Inlet liners, SPME-GC, Supelco®
Kromatografiaan tarvikkeet Kromatografiaan läpivientiholkki
SPME-GC inlet liners are designed for optimum sample introduction for specific injection techniques that are suitable for Agilent, Perkinelmer, Shimadzu, Finnigan, Varian and Thermo scientific equipped with split/splitless inlets.
  • Deactivated (silanized) glass inlet liner for capillary injection ports