Nikkelidikloridihexahydraatii ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.005% Co)

Toimittaja: VWR Chemicals
Vaara

25851.293EA 263 EUR
25851.293
Nikkelidikloridihexahydraatii ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.005% Co)
Nikkelidikloridihexahydraatii
Kaava: NiCl₂·6H₂O
Sulamispiste: 140 °C
Tiheys: 1,84 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149809
CAS-numero: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529

Order Now

Testitulosten erittely

Assay Min. 98 %
pH (20°C; 5 %) 3.5 - 7.0
Ca (Calcium) Max. 0.02 %
Cd (Cadmium) Max. 10 ppm
Co (Cobalt) Max. 50 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 20 ppm
Zn (Zinc) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR