Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody (APC-Cyanine7) [clone: M5/114.15.2]
Toimittaja: Tonbo Biosciences
25-5321-U025 25-5321-U100
25-5321-U025EA 86.3 EUR
25-5321-U025 25-5321-U100
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody (APC-Cyanine7) [clone: M5/114.15.2]
Vasta-aineet
The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages, and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7. The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also beMore Product Information