Peqlab peqGOLD, DNA Ladder, 100 bp
Toimittaja: VWR Chemicals
25-2010EA 81.8 EUR
25-2010
Peqlab peqGOLD, DNA Ladder, 100 bp
Elektroforeesireagenssit Molekyylipainomarkkerit
DNA ladder for quantitative and qualitative analysis of dsDNA.
  • Range: 10 fragments, 100 to 1000 bp in size
  • Highlighted 500 bp band for easy orientation
  • Also available as ready to use mix containing Orange G dye instead of bromophenol blue