Bromivety 48% ACS, Fluka™

Toimittaja: Honeywell Chemicals
Vaara

244260-2.5L 244260-500ML 244260-1L 244260-10L
244260-2.5LEA 361.3 EUR
244260-2.5L 244260-500ML 244260-1L HONC244260-10L
Bromivety 48% ACS, Fluka™
Bromivety
Kaava: HBr
Kiehumispiste: 126 °C (1,013 hPa)
Sulamispiste: -11 °C
Tiheys: 1,490 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011323
CAS-numero: 10035-10-6
UN: 1788
ADR: 8,II

Order Now

Testitulosten erittely

Vapor Pressure 320 psi ( 21.1 °C)
Assay 47.0 - 49.0 %
Residue on ignition (SO₄) Max. 0.002 %
Iron (Fe) Max. 1 ppm
Ignition Residue ≤0.002%
Selenium (Se) Max. 0.01 ppm
Heavy metals (ICP) ACS Max. 5 ppm
Chloride (Cl) Max. 0.05 %
Iodide (I) Max. 0.003 %
Phosphate (PO₄) Max. 0.001 %
Sulfate and sulfite (as SO₄) Max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR