D(+)-glukoosi, anhydrous, GPR RECTAPUR®

Toimittaja: VWR Chemicals

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

24379.294EA 51.8 EUR
24379.294 24379.363 24379.465
D(+)-glukoosi, anhydrous, GPR RECTAPUR®
D(+)-glukoosi
Kaava: C₆H₁₂O₆
MW: 180.16 g/mol
Sulamispiste: 146
Tiheys: 1.54 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00148912
CAS-numero: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Spec. opt. rotation (calc. on anhydrous) 52.5 - 53.3 °
Loss on drying (100°C) Max. 1 %
Ignition residue (SO4) Max. 0.1 %
Cl (Chloride) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR