Rauta(III)kloridi 41% in aqueous solution, TECHNICAL

Toimittaja: VWR Chemicals
Vaara

Synonyms: Iron trichloride, Ferric chloride

24212.298EA 110 EUR
24212.298 24212.367 24212.447
Rauta(III)kloridi 41% in aqueous solution, TECHNICAL
Rauta(III)kloridi
Kaava: FeCl₃
MW: 162.21 g/mol
Kiehumispiste: 315
Sulamispiste: 260
MDL numero: MFCD00011005
CAS-numero: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8 III
Merck Index: 13,04048

Order Now

Testitulosten erittely

Assay 38 - 42 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR