Rauta(III)kloridi 27,5% in aqueous solution, AnalaR NORMAPUR® analytical reagent

Toimittaja: VWR Chemicals
Vaara

Synonyms: Iron trichloride, Ferric chloride

24207.291EA 171 EUR
24207.291
Rauta(III)kloridi 27,5% in aqueous solution, AnalaR NORMAPUR® analytical reagent
Rauta(III)kloridi
Kaava: FeCl₃
MW: 162.21 g/mol
Kiehumispiste: 315
Sulamispiste: 260
MDL numero: MFCD00011005
CAS-numero: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8 III
Merck Index: 13,04048

Order Now

Testitulosten erittely

Assay 27.5 - 29.0 %
Acidity Max. 0.08 meq/g
Density (20/4) 1.260 - 1.280
Free chlorine Max. 10 ppm
Heavy metals (as Pb) Max. 30 ppm
Not precipitated by NH4OH (as SO4) Max. 0.05 %
NH4 (Ammonium) Max. 20 ppm
NO3 (Nitrate) Max. 20 ppm
PO4 (Phosphate) Max. 20 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 5 ppm
Fe (II) (Iron) Max. 30 ppm
Zn (Zinc) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR