Double burette clamps
Toimittaja: Brand
578000
241-0350EA 109 EUR
241-0350
Double burette clamps
Kourat Yleiskäyttöiset puristimet
Aluminium, PVC coated.
  • For two burettes up to 50 ml
  • PVC-coated clamps provide a secure grip
  • Comfortable height adjustment
  • Graduations remain visible
  • Very sturdy design, easy to operate
  • Integral support connector