Petrolieetteri, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)

Toimittaja: VWR Chemicals
Vaara

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

23835.294EA 54.2 EUR
23835.294 23835.328 23835.460 23835.556 23835.363
Petrolieetteri, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
Petrolieetteri
Kiehumispiste: 40…60 °C (1013 hPa)
Tiheys: 0.645…0.665 g/cm³ (20 °C)
Leimahduspiste: –40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081849
CAS-numero: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II
REACH: 01-2119474679-18

Order Now

Testitulosten erittely

Acidity Max. 0.0002 meq/g
Bromine value Max. 1
Colouration Max. 10 APHA
Boiling range (40-60°C) (Vol %) Min. 90%
Substances coloured by H2SO4 Max. 150 APHA
Benzene Max. 100 ppm
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 50 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.2 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.3 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.03 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR