Dietyylieetteri ≥99% stabilised, GPR RECTAPUR® peroxide-free

Toimittaja: VWR Chemicals
Vaara

Synonyms: Eetteri

Stabilization: 4-7 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

23806.328EA 69 EUR
23806.328
Dietyylieetteri ≥99% stabilised, GPR RECTAPUR® peroxide-free
Dietyylieetteri
Kaava: (CH₃CH₂)₂O
MW: 74.12 g/mol
Kiehumispiste: 35
Sulamispiste: –123 °C
Tiheys: 0.7135 g/cm³ (20 °C)
Leimahduspiste: –40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011646
CAS-numero: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay Min. 99 %
Assay (Ionol/BHT) 4 - 10 ppm
Acidity or alkalinity Max. 0.0002 meq/g
Aldehydes + ketones (as HCHO) Max. 5 ppm
Evaporation residue Max. 20 ppm
Peroxides (as H2O2) Max. 0.15 ppm
Water Max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR