Compact safety padlocks
Toimittaja: Brady
814140 814128 814129 814130 814131 814132 814133 814134 814135 814136 814137 814126 814139 814125 814141 814142 814143 814144 814145 814146 814147 814148 814149 814150 814151 814152 814138 814153 814127 814114 814115 814116 814117 814118 814119 814120 814121 814122 814123 814124
238-0108EA 158 EUR
238-0108 238-0096 238-0097 238-0098 238-0099 238-0100 238-0101 238-0102 238-0103 238-0104 238-0105 238-0094 238-0107 238-0093 238-0109 238-0110 238-0111 238-0112 238-0113 238-0114 238-0115 238-0116 238-0117 238-0118 238-0119 238-0120 238-0106 238-0121 238-0095 238-0082 238-0083 238-0084 238-0085 238-0086 238-0087 238-0088 238-0089 238-0090 238-0091 238-0092
Compact safety padlocks
Turvalukot
These padlocks offer the same features as the standard safety padlocks. Though specifically designed for electrical lockout applications, they are smaller and lighter than the standard non conductive and spark resistant padlocks.
  • Reinforced nylon, with aluminium shackle
  • Key will not be released until the padlock shackle is closed
  • 6-pin locking mechanism cylinder (rugged nylon) is ribbed in design for easy handling and grip
  • Nylon casing is chemical and corrosion resistant with enhanced impact resistance
  • Each padlock is keyed differently and is supplied with