4-Amino-N,N-dimetyylianiliini 2 HCl ≥99.0% analytical reagent

Toimittaja: VWR Chemicals
Vaara

Synonyms: N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl

23484.132EA 108 EUR
23484.132
4-Amino-N,N-dimetyylianiliini 2 HCl ≥99.0% analytical reagent
4-Amino-N,N-dimetyylianiliini 2 HCl
Kaava: (CH₃)₂NC₆H₄NH₂·2HCl
MW: 209.12 g/mol
Sulamispiste: 208…212 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012991
CAS-numero: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1 II
Merck Index: 13,03280

Order Now

Testitulosten erittely

Assay Min. 99.0 %
Ignition residue (SO4) Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR