Metyleenikloridi ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent

Toimittaja: VWR Chemicals
Varoitus


Stabilization: Stabiili etanolilla 0,2 %

23354.292EA 29.3 EUR
23354.292 23354.326 23354.550 23354.460 23354.950
Metyleenikloridi ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Metyleenikloridi
Kaava: CH₂Cl₂
Kiehumispiste: 39,8 °C (1013 hPa)
Sulamispiste: –95 °C
Tiheys: 1,322 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000881
CAS-numero: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99.5 %
Acidity Max. 0.0005 meq/g
Boiling point 39.0 - 40.5 °C
Colouration Max. 10 APHA
Density (20/4) 1.322 - 1.327
Substances coloured by H2SO4 Max. 60 APHA
Carbon tetrachloride Max. 100 ppm
Chloroform Max. 100 ppm
Ethanol 0.15 - 0.25 %
Evaporation residue Max. 10 ppm
Free chlorine Max. 0.3 ppm
Water Max. 0.02 %
Al (Aluminium) Max. 0.05 ppm
B (Boron) Max. 0.01 ppm
Ba (Barium) Max. 0.01 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.05 ppm
Zn (Zinc) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR