Forceps, ESD-safe
Toimittaja: OUTILS RUBIS
NK6 NK7 NK2A NKR NK35A
232-2171EA 27.2 EUR
232-2171 232-2172 232-2173 232-2174 232-2175
Forceps, ESD-safe
Pinsetit
Conductive Vestamid with 25% fibre glass reinforcement, black. Precise, robust and virtually unbreakable forceps. Very precise; when pressure is applied during work, the tips don’t open.
  • Electrically conductive
  • Very high chemical resistance to acids and bases
  • Heat resistant up to 175 °C
DIN 53482
Sertifikaatit: DIN 53482