Watchmaker forceps
Toimittaja: MURRAY S
E010/07 E010/04 E010/03 E010/05
231-0435EA 11.8 EUR
231-0435 232-0202 232-0201 232-0203
Watchmaker forceps
Pinsetit
Stainless steel. Ideal for small-scale dissection applications or precise part placement.
  • Very fine points
  • Smooth flat handle
  • Available straight or curved