Kobolttioksidi ≥95%

Toimittaja: Thermo Fisher Scientific
Varoitus

23141.14 23141.22 AB204251.0025
23141.14EA 242 EUR
23141.14 23141.22 ABCRAB204251.0025
Kobolttioksidi ≥95%
Kobolttioksidi

Purity: 95% CAS No.: 1307-96-6 Molecular Formula: CoO Formula Weight: 74.93 Physical Form: Powder Density: 6.45 Appearance: Black-green Melting Point: 1935° Solubility: Soluble in acids or alkalies. Insoluble in water Merck Reference: 14,2446 UN No.: 2811 MDL No.: MFCD00016031


Pigments, glass Colouring, semiconductors

Kaava: CoO
Sulamispiste: 1935 °C
Tiheys: 6,45
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016031
CAS-numero: 1307-96-6
EINECS: 215-154-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,2446

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR