Vibration spatula
Toimittaja: BOCHEM
3540
231-1289EA 36.1 EUR
231-1289
Vibration spatula
Lastat
Stainless steel, plastic shaft. A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material.