Lusikat, näytteenottolusikat, SteriPlast
Toimittaja: BURKLE
5378-1011 5378-1012
231-0525EA 80.9 EUR
231-0525 231-0526
Lusikat, näytteenottolusikat, SteriPlast
Lusikat
PS, white. These disposable spoons are ideal for sampling powders, granules and pastes.
  • Available with capacity of 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
  • Individually packed, gamma radiation sterilised