Spatulas, Trulla type
Toimittaja: NICKEL ELECTRO
9370
231-0393EA 48.8 EUR
231-0393
Spatulas, Trulla type
Lastat
Stainless steel 18/8 non-magnetic, bright finish. Double ended spatulas with trowel shaped scoops for handling large quantities of material.
  • Excellent thermal resistance, autoclavable
  • Excellent chemical resistance
  • Quick and easy to clean