Spatulas, micro, set
Toimittaja: RSG
087.000
231-0195EA 20 EUR
231-0195
Spatulas, micro, set
Lastat
Stainless steel. Set of four 150 mm (overall length) micro spatulas with black plastic handle.
  • Choice of 4 blade widths (3, 4, 5, 6 mm) in one set
  • 45 mm blade length