Rikki 1000 mg/L vedessä (from (NH₄)₂SO₄) Specpure® standard for AAS

Toimittaja: Thermo Fisher Scientific

22958.AE 22958.AP
22958.AEEA 101 EUR
22958.AE 22958.AP
Rikki 1000 mg/L vedessä (from (NH₄)₂SO₄) Specpure® standard for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Sulphur Standards

Molecular Formula: (NH₄)₂SO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00085316


Sertifikaatit: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR