Tubing clips
Toimittaja: UNEX METALLGESELLSCHAFT
10-1204 10-1205 10-1207 10-1210 10-1214 10-1218 10-1222 10-1215
229-7411EA 8.5 EUR
229-7411 229-7412 229-7413 229-7414 229-7415 229-7416 229-7417 229-7418
Tubing clips
Kourat Letkupuristimet
Nickel-chromium steel strip with hexagon head screw.
  • For securing tubes firmly
  • Hexagon head screw for rapid assembly/disassembly
  • Very good chemical and temperature resistance
  • Antimagnetic