Stopcocks, Rotaflo®
Toimittaja: SciLabware
GP3ST/5 GP6ST/7 GP3RA/5 GP6RA/7 HP3ST/5 HP6ST/7 HP10ST/10 HP3RA/5 HP6RA/7 HP10RA/10 HP6RA/10 HP3RA/7 HP3ST/7 GP10ST/10 HP6ST/10
201-8222EA 368 EUR
201-8222 201-8224 201-8223 201-8225 201-8231 201-8232 201-8233 201-8234 201-8235 229-0232 229-0233 BIBBHP3RA/7 229-0242 BIBBGP10ST/10 BIBBHP6ST/10
Stopcocks, Rotaflo®
Hanat
PYREX®, borosilicate glass with PTFE needle valve.
  • Lubricant is not required (no seizure, no contamination with grease)
  • Interchangeable elements
  • Precise flow control
These chemically inert stopcocks can be used at temperatures up to +200 °C. Two different models are available: general purpose (vacuum rating up to 10-4 mm Hg) and high performance (vacuum rating up to 10-9 mm Hg).