Tubing, silicone, EXACANAL
Toimittaja: Saint Gobain Life Sciences
760350 760180
228-1607EA 196 EUR
228-1607 228-1194
Tubing, silicone, EXACANAL
Letku Siirtoputki
Silicone COVOSIL 9161, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Very flexible
Temperature resistance: –60 to +200 °C
Hardness (Shore durometer A): 60
Density: 1,14
Sertifikaatit: Complies with FDA (21 CFR § 177.2600) standards